Anti-CHREBP Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NB400-135G
102131-520EA 675.69 USD
102131-520
Anti-CHREBP Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The CHREBP Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to CHREBP. This antibody reacts with human, mouse, rat. The CHREBP Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CHREBP
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR