Anti-Choline Acetyltransferase/ChAT Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-89724
102126-158EA 564.74 USD
102126-158
Anti-Choline Acetyltransferase/ChAT Rabbit Polyclonal Antibody
Antibodies
The Choline Acetyltransferase / ChAT Antibody from Novus Biologicals is a rabbit polyclonal antibody to Choline Acetyltransferase / ChAT. This antibody reacts with mouse, rat. The Choline Acetyltransferase / ChAT Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:300-1:2000

Type: Primary
Antigen: Choline Acetyltransferase/ChAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR