Anti-CENPF Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB500-101R
102131-778EA 379 USD
102131-778
Anti-CENPF Rabbit Polyclonal Antibody (DyLight 550)
The CENPF Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to CENPF. This antibody reacts with human, bovine. The CENPF Antibody [DyLight 550] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CENPF
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR