Anti-CELSR2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NLS1942
102253-164EA 329 USD
102253-164
Anti-CELSR2 Rabbit Polyclonal Antibody
Antibodies
The CELSR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CELSR2. This antibody reacts with human. The CELSR2 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 15-20 ?g/ml

Type: Primary
Antigen: CELSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR