Anti-Cdk9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB120-10874
102126-766EA 635.51 USD
102126-766
Anti-Cdk9 Rabbit Polyclonal Antibody
Antibodies
The Cdk9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cdk9. This antibody reacts with human, mouse, rat. The Cdk9 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Cdk9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR