Anti-VEGF-D Mouse Monoclonal Antibody [clone: MM0007-7E79]

Supplier: Novus Biologicals
NB110-60973
102124-316EA 635.51 USD
102124-316
Anti-VEGF-D Mouse Monoclonal Antibody [clone: MM0007-7E79]
Antibodies
The VEGF-D Antibody (MM0007-7E79) from Novus Biologicals is a mouse monoclonal antibody to VEGF-D. This antibody reacts with human. The VEGF-D Antibody (MM0007-7E79) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200-1:500

Type: Primary
Antigen: VEGF-D
Clonality: Monoclonal
Clone: MM0007-7E79
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR