Anti-TWEAK/TNFSF12 Mouse Monoclonal Antibody [clone: CARL-1]

Supplier: Novus Biologicals
NB100-77867
102117-432EA 199 USD
102117-432
Anti-TWEAK/TNFSF12 Mouse Monoclonal Antibody [clone: CARL-1]
The TWEAK / TNFSF12 Antibody (CARL-1) from Novus Biologicals is a mouse monoclonal antibody to TWEAK / TNFSF12. This antibody reacts with human. The TWEAK / TNFSF12 Antibody (CARL-1) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TWEAK/TNFSF12
Clonality: Monoclonal
Clone: CARL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR