Anti-TRF-1 Mouse Monoclonal Antibody (Biotin) [clone: 57-6]

Supplier: Novus Biologicals
NB110-68281B
102124-690EA 614.15 USD
102124-690
Anti-TRF-1 Mouse Monoclonal Antibody (Biotin) [clone: 57-6]
Antibodies
The TRF-1 Antibody (57-6) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to TRF-1. This antibody reacts with human, mouse, rat, monkey. The TRF-1 Antibody (57-6) [Biotin] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TRF-1
Clonality: Monoclonal
Clone: 57-6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR