Anti-Serum Amyloid A1 Mouse Monoclonal Antibody [clone: SAA15]

Supplier: Novus Biologicals
NB100-73077
102115-932EA 417.23 USD
102115-932
Anti-Serum Amyloid A1 Mouse Monoclonal Antibody [clone: SAA15]
Antibodies
The Serum Amyloid A1 Antibody (SAA15) from Novus Biologicals is a mouse monoclonal antibody to Serum Amyloid A1. This antibody reacts with human. The Serum Amyloid A1 Antibody (SAA15) has been validated for the following applications: Western Blot, Sandwich ELISA.

Type: Primary
Antigen: Serum Amyloid A1
Clonality: Monoclonal
Clone: SAA15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR