Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S-arrestin Mouse Monoclonal Antibody (DyLight 650) [clone: PDS-1]

Supplier: Novus Biologicals
NB100-2385C
102107-312
Anti-S-arrestin Mouse Monoclonal Antibody (DyLight 650) [clone: PDS-1]
The S-arrestin Antibody (PDS-1) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to S-arrestin. This antibody reacts with human, porcine, bovine. The S-arrestin Antibody (PDS-1) [DyLight 650] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: S-arrestin
Clonality: Monoclonal
Clone: PDS-1
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine, Bovine

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty