Anti-rRNA Mouse Monoclonal Antibody (Biotin) [clone: Y10b]

Supplier: Novus Biologicals
NB100-662B
102115-240EA 700.31 USD
102115-240
Anti-rRNA Mouse Monoclonal Antibody (Biotin) [clone: Y10b]
Antibodies
The rRNA Antibody (Y10b) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to rRNA. This antibody reacts with human, mouse, all species. The rRNA Antibody (Y10b) [Biotin] has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: rRNA
Clonality: Monoclonal
Clone: Y10b
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse, All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR