Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: 9C5]

Supplier: Novus Biologicals
NB110-7545
102125-290EA 380.31 USD
102125-290
Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: 9C5]
Antibodies
The Respiratory Syncytial Virus Antibody (9C5) from Novus Biologicals is a mouse monoclonal antibody to Respiratory Syncytial Virus. This antibody reacts with virus. The Respiratory Syncytial Virus Antibody (9C5) has been validated for the following applications: ELISA.

Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: Monoclonal
Clone: 9C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR