Anti-GH1 Mouse Monoclonal Antibody [clone: GH-45]

Supplier: Novus Biologicals
NB500-364
102132-320EA 564.74 USD
102132-320
Anti-GH1 Mouse Monoclonal Antibody [clone: GH-45]
Antibodies
Mouse Monoclonal Growth Hormone Antibody (GH-45). Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR