Anti-COX-2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-868
102118-720EA 564.74 USD
102118-720
Anti-COX-2 Goat Polyclonal Antibody
Antibodies
The COX-2 Antibody from Novus Biologicals is a goat polyclonal antibody to COX-2. This antibody reacts with human, mouse. The COX-2 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR