Anti-IgG Donkey Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Novus Biologicals
NB7590
102135-418EA 437.35 USD
102135-418
Anti-IgG Donkey Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Donkey Polyclonal Donkey anti-Goat IgG Antibody [R-Phycoerythrin]. Tested Applications: Immunocytochemistry/Immunofluorescence. Tested Reactivity: Goat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR