Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Novus Biologicals
NB7356
102135-182EA 295.82 USD
102135-182
Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Donkey Polyclonal Donkey anti-Goat IgG Antibody [FITC]. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Goat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:2

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR