Anti-Beclin 1 Antibody

Supplier: Novus Biologicals
NB910-95610
102136-980EA 848 USD
102136-980
Anti-Beclin 1 Antibody
Antibodies
The Beclin 1 Antibody Pack from Novus Biologicals is a set of antibodies to Beclin 1. These antibodies react with human, mouse, rat. The Beclin 1 Antibody Pack has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Antibody Packs
Antigen: Beclin 1
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR