Anti-Autophagy Antibody

Supplier: Novus Biologicals
NB910-94159
102136-976EA 774.15 USD
102136-976
Anti-Autophagy Antibody
Antibodies
The Autophagy Antibody Pack from Novus Biologicals is a set of antibodies to Autophagy. These antibodies react with human, mouse. The Autophagy Antibody Pack has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Antibody Packs
Antigen: Autophagy
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR