Anti-TNFA Armenian Hamster Monoclonal Antibody

NB110-9916
102126-510EA 466.15 USD
102126-510
Anti-TNFA Armenian Hamster Monoclonal Antibody
Armenian Hamster Monoclonal TNF alpha Antibody (TN3-19.12). Tested Applications: Western Blot, Blocking/Neutralizing, ELISA, Immunoprecipitation. Tested Reactivity: Mouse, Rat.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR