Anti-NUP62 Armenian Hamster Monoclonal Antibody [clone: RL31]

Supplier: Novus Biologicals
NB120-2736
102127-610EA 564.74 USD
102127-610
Anti-NUP62 Armenian Hamster Monoclonal Antibody [clone: RL31]
Antibodies
The NUP62 Antibody (RL31) from Novus Biologicals is a armenian hamster monoclonal antibody to NUP62. This antibody reacts with rat. The NUP62 Antibody (RL31) has been validated for the following applications: Western Blot, Immunofluorescence.

Type: Primary
Antigen: NUP62
Clonality: Monoclonal
Clone: RL31
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR