Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Armenian Hamster Monoclonal anti-Integrin alpha E Antibody (2E7) [Biotin], Novus Biologicals (NBP1-28125)

Supplier: Novus Biologicals
NBP1-28125
Armenian Hamster Monoclonal Integrin alpha E Antibody (2E7) [Biotin]. Tested Applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Immunoprecipitation. Tested Reactivity: Mouse.

Type: Primary
Antigen: Integrin alpha E
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Hamster (American)
Isotype:
Reactivity: Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.25 mg NBP1-28125 102161-620 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ITGAE
Antigen Name Integrin alpha E
Reactivity Mouse
Conjugation Biotin
Clonality Monoclonal
Antibody Type Primary
Clone 20000000
Host Armenian Hamster
Isotype IgG2a
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms Mucosal lymphocyte 1 antigen, integrin alpha-E, human mucosal lymphocyte antigen 1, alpha E (antigen CD103, CD103 antigen, Integrin alpha-IEL, alphapolypeptide), antigen CD103, alpha polypeptide, CD103, HML-1 antigen, HUMINAE, integrin, MGC141996
Gene ID 3682
Concentration 0.5 mg/ml
Immunogen C57BL/6J mouse intestinal intraepithelial lymphocytes (IEL)
Purification Protein A or G purified
Storage Buffer PBS
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty