Anti-IL10RA Armenian Hamster Monoclonal Antibody [clone: RM0101-9E12]

Supplier: Novus Biologicals
NBP1-22459
102160-442EA 635.51 USD
102160-442
Anti-IL10RA Armenian Hamster Monoclonal Antibody [clone: RM0101-9E12]
Antibodies
Armenian Hamster Monoclonal IL10RA Antibody (RM0101-9E12). Tested Applications: Western Blot. Tested Reactivity: Mouse.

Type: Primary
Antigen: IL10RA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR