Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EMR3 Armenian Hamster Monoclonal Antibody (Unconjugated) [clone: 3D7]

Supplier: Novus Biologicals
NB100-65935
The EMR3 Antibody (3D7) from Novus Biologicals is a armenian hamster monoclonal antibody to EMR3. This antibody reacts with human. The EMR3 Antibody (3D7) has been validated for the following applications: Flow Cytometry, Immunoprecipitation.

Type: Primary
Antigen: EMR3
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NB100-65935 102115-112 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol EMR3
Antigen Name EGF-like module Containing mucin-like Hormone Receptor-like 3
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 3D7
Host Armenian Hamster
Isotype IgG
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms egf-like module-containing mucin-like receptor 3, EGF-like module receptor 3, egf-like module containing, hormone receptor-like 3, EGF-like module-containing mucin-like hormone receptor-like 3, mucin-like
Gene ID 84658
Concentration 1.0 mg/ml
Immunogen ARHO-EMR3-CD97 (EGF1) transfectants.
Purification Protein G purified
Storage Buffer PBS (pH 7.4)
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty