Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTLA-4 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1B8]

Supplier: Novus Biologicals
NBP1-28066
The CTLA-4 Antibody (1B8) [FITC] from Novus Biologicals is a armenian hamster monoclonal antibody to CTLA-4. This antibody reacts with mouse. The CTLA-4 Antibody (1B8) [FITC] has been validated for the following applications: Flow Cytometry, In vitro assay.

Type: Primary
Antigen: CTLA-4
Clonality: Monoclonal
Clone: 1B8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.25 mg NBP1-28066 102161-536 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CTLA-4
Antigen Name Cytotoxic T-lymphocyte-Associated Protein 4
Reactivity Mouse
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Monoclonal
Antibody Type Primary
Clone 1B8
Host Armenian Hamster
Isotype IgG
Flow Cytometry Yes
Antigen Synonyms cytotoxic T-lymphocyte-associated protein 4, CD28, ICOS, cytotoxic T-lymphocyte antigen 4, GRD4, CD152 antigen, CELIAC3, cytotoxic T-lymphocyte protein 4, GSE, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4, CD152IDDM12, cytotoxic T-lymphocyte-associated serine esterase-4, celiac disease 3, Cytotoxic T-lymphocyte-associated antigen 4, CD, CTLA-4
Gene ID 1493
Immunogen Extracellular portion of mouse CD152 fused to a mouse IgG2a.
Purification Protein A or G purified
Storage Buffer PBS
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty