Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD79B Armenian Hamster Monoclonal Antibody (Unconjugated) [clone: HM79-11]

Supplier: Novus Biologicals
The CD79B Antibody (HM79-11) from Novus Biologicals is a armenian hamster monoclonal antibody to CD79B. This antibody reacts with mouse. The CD79B Antibody (HM79-11) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: HM79-11
Conjugation: Unconjugated
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NB100-65611 102114-880 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol CD79B
Antigen Name Cluster of Differentiation 79a molecule, immunoglobulin-associated beta
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone HM79-11
Host Armenian Hamster
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms Immunoglobulin-associated B29 protein, immunoglobulin-associated beta, CD79b antigen, Ig-beta, CD79b molecule, B29B-cell-specific glycoprotein B29, IGBAGM6, B-cell antigen receptor complex-associated protein beta chain, CD79B antigen (immunoglobulin-associated beta)
Gene ID 974
Concentration 1.0 mg/ml
Immunogen CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells.
Purification Protein G purified
Storage Buffer PBS (pH 7.4)
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty