Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD69 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H1.2F3]

Supplier: Novus Biologicals
NBP1-28012
The CD69 Antibody (H1.2F3) [FITC] from Novus Biologicals is a armenian hamster monoclonal antibody to CD69. This antibody reacts with mouse. The CD69 Antibody (H1.2F3) [FITC] has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG1
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.25 mg NBP1-28012 102161-456 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CD69
Antigen Name CD69 molecule
Reactivity Mouse
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Monoclonal
Antibody Type Primary
Clone H1.2F3
Host Armenian Hamster
Isotype IgG1
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms Early T-cell activation antigen p60, EA1, CD69 molecule, early activation antigen CD69, activation inducer molecule (AIM/CD69), BL-AC/P26, C-type lectin domain family 2, Activation inducer molecule, early T-cell activation antigen), member C, CD69 antigen (p60, MLR-3, CD69 antigen, CLEC2CC-type lectin domain family 2 member C, early lymphocyte activation antigen, GP32/28, Leukocyte surface antigen Leu-23, AIM
Gene ID 969
Immunogen Mouse dendritic epidermal cell line Y245
Purification Protein A or G purified
Storage Buffer PBS
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty