3-(Pentafluorothio)aniline ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Aminophenylsulphur pentafluoride
066645-1G
066645-5G
101780-834EA
83.08
USD
101780-834
101780-836
3-(Pentafluorothio)aniline ≥95%
3-(Pentafluorothio)aniline
Hazard: Toxic/Irritant
Formula:
C₆H₆F₅NS MW: 218.17 Melting Pt: 35 °C |
MDL Number:
MFCD00674199 CAS Number: 2993-22-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...