2-Nitroimidazole

Supplier: Matrix Scientific

Synonyms: 2-Nitro-1H-imidazole

065643-1G 065643-5G
101780-828EA 23.08 USD
101780-828 101780-830
2-Nitroimidazole
2-Nitroimidazole

Hazard: Toxic

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: ∼. 287 °C (dec.)
MDL Number: MFCD00005185
CAS Number: 527-73-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR