(3-Bromo-4-pyridyl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-pyridinemethanol3-Bromo-4-(hydroxymethyl)pyridine

064048-1G
101780-826EA 113.85 USD
101780-826
(3-Bromo-4-pyridyl)methanol ≥97%
(3-Bromo-4-pyridyl)methanol

Hazard: Flammable Solid/Irritant

Formula: C₆H₆BrNO
MW: 188.03 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD11847285
CAS Number: 146679-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR