Cyclopropyl isothiocyanate

Supplier: Matrix Scientific

063222-500MG
101780-820EA 183.08 USD
101780-820
Cyclopropyl isothiocyanate
Cyclopropyl isothiocyanate

Hazard: Flammable/Toxic/Moisture Sensitive/Refrigerate

Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 87…88 °C (35 mmHg)
Density: 1.07
Flash Pt: 57 °C
MDL Number: MFCD00040874
CAS Number: 56601-42-4
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR