2,3,6-Trifluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

059601-5G 059601-1G
101780-796EA 158.46 USD
101780-796 101780-794
2,3,6-Trifluorobenzenesulfonyl chloride ≥97%
2,3,6-Trifluorobenzenesulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₂ClF₃O₂S
MDL Number: MFCD09832391
CAS Number: 1017779-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR