3-Cyano-4-fluorobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

058558-1G
101780-780EA 170.77 USD
101780-780
3-Cyano-4-fluorobenzenesulfonamide ≥97%
3-Cyano-4-fluorobenzenesulfonamide

Hazard: Toxic

Formula: C₇H₅FN₂O₂S
Melting Pt: 150…153 °C
MDL Number: MFCD16705414
CAS Number: 928139-30-4

Order Now

Specification Test Results

Melting point 150-153°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR