Benzyloxyacetyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Benzyloxy)acetyl chloride

057597-25G 057597-5G
101780-764EA 72.31 USD
101780-764 101780-766
Benzyloxyacetyl chloride ≥95%
Benzyloxyacetyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 84…87 °C (0.4 mmHg)
Density: 1.189
MDL Number: MFCD00010768
CAS Number: 19810-31-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 84-87°/0.4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR