2-Methoxybenzyl chloride ≥98%

057115-250G
101780-758EA 835.87 USD
101780-758
2-Methoxybenzyl chloride ≥98%
2-Methoxybenzyl chloride

50% by weight in Methylene chloride


Hazard: Corrosive/Refrigerate

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 85 °C (1 mmHg)
Melting Pt: 28…29 °C
Density: 1.125
Flash Pt: 210 °F
CAS Number: 7035-02-1

Order Now

Specification Test Results

Boiling point 85°/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR