3-Chloro-2-methylanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-3-methoxy-2-methylbenzene2-Chloro-6-methoxytoluene

056960-5G
101780-708EA 107.69 USD
101780-708
3-Chloro-2-methylanisole ≥98%
3-Chloro-2-methylanisole

Hazard: Flammable

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 213…217 °C
Density: 1.14
Flash Pt: 92 °C
MDL Number: MFCD00070772
CAS Number: 3260-88-6

Order Now

Specification Test Results

Boiling point 213-217°
Density 1.14

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR