4-tert-Butylbenzylamine
Supplier: Matrix Scientific
Synonyms:
p-tert-Butylbenzylamine
056939-25G
056939-5G
101780-698EA
106.15
USD
101780-698
101780-700
4-tert-Butylbenzylamine
4-tert-Butylbenzylamine
Hazard: Corrosive
Formula:
C₁₁H₁₇N MW: 163.26 g/mol Boiling Pt: 69 °C (1 mmHg) Density: 0.93 |
MDL Number:
MFCD00040754 CAS Number: 39895-55-1 |
Specification Test Results
Boiling point | 69°/1mm |
Density | 0.93 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...