6-Chloro-2-hydroxy-3-nitropyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Chloro-3-nitropyridin-2(1H)-one
054608-5G
101780-662EA
90.77
USD
101780-662
6-Chloro-2-hydroxy-3-nitropyridine ≥97%
6-Chloro-2-hydroxy-3-nitropyridine
Hazard: Toxic/Irritant
Formula:
C₅H₃ClN₂O₃ MW: 174.54 g/mol |
MDL Number:
MFCD05261002 CAS Number: 92138-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...