7-Nitro-2,3-dihydro-1-benzofuran-5-sulfonoylchloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3-Dihydro-7-nitrobenzo[b]furan-5-sulphonyl chloride
047060-5G
047060-1G
047060-500MG
101780-628EA
923.08
USD
101780-628
101780-624
101780-626
7-Nitro-2,3-dihydro-1-benzofuran-5-sulfonoylchloride ≥95%
7-Nitro-2,3-dihydro-1-benzofuran-5-sulfonoylchloride
Hazard: Corrosive
Formula:
C₈H₆ClNO₅S MW: 263.66 g/mol Melting Pt: 111…113 °C |
MDL Number:
MFCD03408011 CAS Number: 874781-18-7 |
Specification Test Results
Melting point | 111-113° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...