2-Chloro-4-isocyanato-1-methoxybenzene

Supplier: Matrix Scientific

038707-500MG
101780-620EA 221.54 USD
101780-620
2-Chloro-4-isocyanato-1-methoxybenzene
2-Chloro-4-isocyanato-1-methoxybenzene

Hazard: Toxic/Lachrymator

Formula: C₈H₆ClNO₂
Melting Pt: 30…33 °C
MDL Number: MFCD02093651
CAS Number: 28395-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR