1-Isocyanato-3-methylbutane

Supplier: Matrix Scientific

038688-500MG
101780-614EA 290.77 USD
101780-614
1-Isocyanato-3-methylbutane
1-Isocyanato-3-methylbutane

Hazard: Flammable/Toxic

Formula: C₆H₁₁NO
Boiling Pt: 130 °C (760 mmHg)
MDL Number: MFCD09901277
CAS Number: 1611-65-0

Order Now

Specification Test Results

Boiling point 130°/760mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR