Methyl-3-isocyanatopropanoate
Supplier: Matrix Scientific
038033-1G
038033-5G
101780-594EA
136.92
USD
101780-594
101780-596
Methyl-3-isocyanatopropanoate
Methyl-3-isocyanatopropanoate
Hazard: Toxic/Irritant
Formula:
C₅H₇NO₃ MW: 129.12 g/mol |
MDL Number:
MFCD00797846 CAS Number: 50835-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...