2,5-Dichloro-4-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

035667-1G
101780-576EA 223.08 USD
101780-576
2,5-Dichloro-4-(trifluoromethyl)pyridine ≥97%
2,5-Dichloro-4-(trifluoromethyl)pyridine

Hazard: Flammable/Irritant-Harmful

Formula: C₆H₂Cl₂F₃N
MDL Number: MFCD09749663
CAS Number: 89719-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR