2-(Trifluoromethyl)pyrazine ≥97%

Supplier: Matrix Scientific

034762-1G
101780-568EA 136.85 USD
101780-568
2-(Trifluoromethyl)pyrazine ≥97%
2-(Trifluoromethyl)pyrazine

Hazard: Toxic

Formula: C₅H₃F₃N₂
MW: 148.09 g/mol
Boiling Pt: 116…118 °C
MDL Number: MFCD06797734
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 116-118°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR