6-(Trifluoromethyl)pyridine-3-sulfonyl chloride ≥97%

Supplier: Matrix Scientific

034759-1G
101780-566EA 213.85 USD
101780-566
6-(Trifluoromethyl)pyridine-3-sulfonyl chloride ≥97%
6-(Trifluoromethyl)pyridine-3-sulfonyl chloride

Hazard: Corrosive/Irritant/Refrigerate

Formula: C₆H₃ClF₃NO₂S
MW: 245.61 g/mol
Melting Pt: 42…45 °C
Flash Pt: >110 °C
MDL Number: MFCD10699112
CAS Number: 959996-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR