6-(Trifluoromethyl)pyridine-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

034740-1G
101780-564EA 210.77 USD
101780-564
6-(Trifluoromethyl)pyridine-2-carbonyl chloride ≥97%
6-(Trifluoromethyl)pyridine-2-carbonyl chloride

Hazard: Corrosive/Irritant/Store in Freezer

Formula: C₇H₃ClF₃NO
Boiling Pt: 88 °C (6 mmHg)
MDL Number: MFCD11226641
CAS Number: 1099597-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR