3-Amino-4-methylpyridine ≥98%

Supplier: Matrix Scientific

034729-25G
101780-552EA 36.92 USD
101780-552
3-Amino-4-methylpyridine ≥98%
3-Amino-4-methylpyridine

Hazard: Toxic

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 254 °C
Melting Pt: 102…106 °C
MDL Number: MFCD00128871
CAS Number: 3430-27-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 254°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR