5-(Trifluoromethyl)pyridine-2-carbonyl chloride ≥95%

Supplier: Matrix Scientific

032537-1G
101780-544EA 476.92 USD
101780-544
5-(Trifluoromethyl)pyridine-2-carbonyl chloride ≥95%
5-(Trifluoromethyl)pyridine-2-carbonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₇H₃ClF₃NO
Boiling Pt: 66…68 °C (6 mmHg)
Density: 1.468
MDL Number: MFCD11226644
CAS Number: 128073-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR