2,4-Dichloropyrimidine-5-carbonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-5-pyrimidinecarbonyl chloride

032528-1G
101780-538EA 158.46 USD
101780-538
2,4-Dichloropyrimidine-5-carbonyl chloride ≥97%
2,4-Dichloropyrimidine-5-carbonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₅HCl₃N₂O
MW: 211.44 g/mol
Boiling Pt: 67…70 °C (0.1 mmHg)
MDL Number: MFCD09742432
CAS Number: 2972-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR