3,6-Difluoro-2-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

032383-1G
101780-526EA 90.77 USD
101780-526
3,6-Difluoro-2-(trifluoromethyl)pyridine ≥95%
3,6-Difluoro-2-(trifluoromethyl)pyridine

Hazard: Toxic

Formula: C₆H₂F₅N
Boiling Pt: 139…142 °C
MDL Number: MFCD10699122
CAS Number: 1099597-92-8

Order Now

Specification Test Results

Boiling point 139-142°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR