2-(Isocyanatomethyl)tetrahydrofuran

Supplier: Matrix Scientific

030196-500MG
101780-512EA 384.62 USD
101780-512
2-(Isocyanatomethyl)tetrahydrofuran
2-(Isocyanatomethyl)tetrahydrofuran

Hazard: Toxic/Irritant

Formula: C₆H₉NO₂
Boiling Pt: 195 °C (760 mmHg)
MDL Number: MFCD09901270
CAS Number: 51998-05-1

Order Now

Specification Test Results

Boiling point 195°/760mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR